missing translation for 'onlineSavingsMsg'
Learn More

Myristic Acid, Sigma-Aldrich™

Product Code. 15898250
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898250 100 g 100g
15808260 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15898250 Supplier Sigma Aldrich Supplier No. 8003990100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-63-8

Specifications

Melting Point 52°C to 56°C
Density 0.8622 g/cm3 at 54°C
Boiling Point 326°C/1013 hPa
Molecular Formula CH3(CH2)12COOH
MDL Number MFCD00002744
Quantity 100 g
Synonym Myristic acid, Tetradecanoic acid, Myristic acid
Molecular Weight (g/mol) 228.37
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.