missing translation for 'onlineSavingsMsg'
Learn More

Myristic acid, 95%, MP Biomedicals™

Product Code. 11443620 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443620 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443620 Supplier MP Biomedicals Supplier No. 0210237583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 544-63-8
Color Tan
Molecular Formula C14H28O2
MDL Number MFCD00002744
Quantity 250 g
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC(O)=O
IUPAC Name tetradecanoic acid
Molecular Weight (g/mol) 228.38
PubChem CID 11005
ChEBI CHEBI:28875
Formula Weight 228.37
Percent Purity 0.95
Physical Form Powder
Chemical Name or Material Myristic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.