Learn More
myo-Inositol, Millipore™
Product Code. 10590771
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10590771 250 g 250g
10070930 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10590771 Supplier Millipore Supplier No. 1045070250
 Restricted Item
Estimated Shipment: 19-11-2026
For biochemistry
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87-89-8
Specifications
Melting Point 225°C to 227°C
MDL Number MFCD00077932
Quantity 250 g
Synonym myo-Inositol, Hexahydroxycyclohexane, Cyclohexanehexol, meso-Inositol, Myo-Inosit, myo-Inositol
Molecular Weight (g/mol) 180.16
Grade Research