Learn More
Novus Biologicals™ MyD88 Inhibitor Peptide Set
Product Code. 18766713 Shop All Bio Techne Products
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Product Code. 18766713
Brand: Novus Biologicals™ NBP229328
Applications: Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 2 mg
For Research Use Only