Learn More
Lonza™ MycoAlert™ Mycoplasma Detection Kit (10 Tests)
Product Code. 11690251
Change view
Click to view available options
Quantity:
2 x 600 μL MycoAlert Reagent, 2 x 600 μL MycoAlert Substrate, 1 x 10 mL MycoAlert Assay Buffer
Unit Size:
10 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11690251 2 x 600 μL MycoAlert Reagent, 2 x 600 μL MycoAlert Substrate, 1 x 10 mL MycoAlert Assay Buffer 10 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11690251 Supplier Lonza™ Supplier No. LT07118
  • ONLINE EXCLUSIVE PRICE
    452.00 EUR
    383.53 EUR / 10 tests
    Save 68.47 EUR
    15% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Selective biochemical test that exploits the activity of mycoplasmal enzymes that are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.
  • Convenient and fast.
  • Simply add two reagents to the culture supernatant.
  • Perform two luminescence readings.
  • Results in less than 20 min.
  • Easy interpretation of results .
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma).
  • Negligible interference with media components.
  • Selective biochemical test that exploits the activity of mycoplasmal enzymes that are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.
  • Viable mycoplasma in a test sample (cell supernatant) are lysed and the enzymes react with the MycoAlert™ Substrate, catalyzing the conversion of ADP to ATP.
  • The ATP is then transferred into a light signal via the luciferase enzyme in the MycoAlert™ Reagent.
  • By measuring the level of ATP in a sample both before (read A) and after the addition of the MycoAlert™ Substrate (read B), a ratio can be obtained which is indicative of the presence or absence of mycoplasma.
TRUSTED_SUSTAINABILITY
Specifications
Color Light yellow
Content And Storage MycoAlert Reagent (lyophilized), MycoAlert Substrate (lyophilized), MycoAlert Assay Buffer
For Use With (Application) Mycoplasma detection
Quantity 2 x 600 μL MycoAlert Reagent, 2 x 600 μL MycoAlert Substrate, 1 x 10 mL MycoAlert Assay Buffer