Learn More
Lonza™ Mycoalert™ Mycoplasma Assay Control Set
Product Code. 11680271
Change view
Click to view available options
Quantity:
10 Tests
Unit Size:
10 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11680271 10 Tests 10 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11680271 Supplier Lonza™ Supplier No. LT07518
 Restricted Item
Estimated Shipment: 16-11-2026
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
It is recommended to always include the Control Set to ensure that the operator and all of the assay reagents are performing properly. Mycoalert™ Mycoplasma Assay Control Set is designed for use with the MycoAlert™ Mycoplasma Detection Kit.
Convenient & fast
  • Simply add two reagents to the culture supernatant
  • Perform two luminescence readings
  • Results in < 20 min
  • Easy interpretation of results
Universal
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Negligible interference with media components
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Mycoalert assay control : 1 x 1mL, Mycoalert assay buffer: 1 x 2mL
2 to 8°C
Cell Type Mammalian Cells
Product Type Mycoplasma Detection
For Use With (Application) Mycoplasma detection
Quantity 10 Tests
Product Identifier
  • MycoAlert Assay Control Set
Hazard Statement
  • EUH210-Safety data sheet available on request.