Learn More
Epredia™ MX35 Premier™ Disposable Low-profile Microtome Blades
Product Code. 12074088 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Microtome Blade, Premier
Microtome Blade, Premier Plus
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12074088 Microtome Blade, Premier Pack of 50
10687875 Microtome Blade, Premier Plus Pack of 50
10454379 Microtome Blade, Premier Plus Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12074088 Supplier Epredia™ Supplier No. 3051835
  • ONLINE EXCLUSIVE PRICE
    161.25 EUR
    80.83 EUR / Pack of 50
    Save 80.42 EUR
    49% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Section hard tissues and high-quality thin sections with high-performance with Epredia™ MX35 Premier™ disposable low-profile microtome blades.
  • Sharper edge with increased durability for longer blade life
  • Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microtome Blade, Premier
Profile Low
For Use With (Application) Designed for routine histology and are good for a variety of tissue types, including hard tissue
Disposable Single-use
Cutting Angle 34°
Thickness (English) 0.01 in.
Thickness (Metric) 0.24 mm