Learn More
Muscimol, TRC
Product Code. 30137963
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137963 25 mg 25mg
18071810 5 mg 5mg
18013620 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137963 Supplier Toronto Research Chemicals Supplier No. TRCM81500025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2763-96-4
Molecular Formula C4 H6 N2 O2
Molecular Weight (g/mol) 114.1
IUPAC Name 5-(aminomethyl)-1,2-oxazol-3-one
SMILES NCC1=CC(=O)NO1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C4H6N2O2/c5-2-3-1-4(7)6-8-3/h1H,2,5H2,(H,6,7)
Formula Weight 114.0429
Percent Purity >95%
Chemical Name or Material Muscimol
Grade HPLC