missing translation for 'onlineSavingsMsg'
Learn More

Murexide, Supelco™

Product Code. 15817440
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817440 5 g 5g
15804780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15817440 Supplier Supelco Supplier No. 1061610005
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ammonium purpurate metal indicator ACS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3051-09-0

Specifications

MDL Number MFCD00012777
Synonym Murexide, Ammonium purpurate, acid, Murexide
Quantity 5 g
Molecular Weight (g/mol) 284.19
Grade Research
Chemical Name or Material Murexide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.