missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin, TRC

Product Code. 30003731
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003731 100 mg 100mg
30004562 10 mg 10mg
30009333 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003731 Supplier Toronto Research Chemicals Supplier No. TRCM794000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12650-69-0
Molecular Formula C26 H44 O9
Molecular Weight (g/mol) 500.62
Synonym L-talo-Non-2-enonic acid, 5,9-anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-, 8-carboxyoctyl ester, [2E,8[2S,3S(1S,2S)]]-, BRL 4910A, Bactoderm, Bactroban, Bactroban Ointment, Mupirocin, Mupirocin Neo-Sensitabs, Mupricin, Pseudomonic acid, Pseudomonic acid A, Turixin, trans-Pseudomonic acid
IUPAC Name 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
SMILES C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)OCCCCCCCCC(=O)O)\C)[C@H](O)[C@@H]2O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H44O9/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29)/b16-13+/t17-,18-,19-,20-,21-,24+,25-,26-/m0/s1
Formula Weight 500.2985
Chemical Name or Material Mupirocin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.