Learn More
ZVG™ Multitex™ Multi-Purpose Wipe
Product Code. 10712544
Change view
Click to view available options
Dimensions (L x W):
38 x 34 cm
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10712544 38 x 34 cm Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10712544 Supplier ZVG™ Supplier No. 1014901
  • ONLINE EXCLUSIVE PRICE
    61.10 EUR
    58.89 EUR / Pack of 400
    Save 2.21 EUR
    3% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Viscose fleece cleaning cloth
  • The high-quality fleeces with textile character are due to their enormous strength for wet and dry cleaning
  • They are very soft and yet lint-free, can absorb large quantities of liquid and remain tear-resistant, are solvent-resistant and food-safe
  • They are regularly used, especially in sensitive work areas such as the precision mechanics, the electrical and vehicle industries and in paint shops
  • Multitex™ is available in different versions: Multitex™ soft is suitable for the treatment of sensitive surfaces due to its fine hole structure, as liquids are absorbed and released evenly. Multitex™ extra can absorb large amounts of liquid thanks to its coarse hole structure.
Specifications
Color Blue
Dimensions (L x W) 38 x 34 cm
Packaging Quantity 400/Pk.