Learn More
Gilson™ Multiple Column Adapter
Product Code. 16666145 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
Fraction Collector
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16666145 Fraction Collector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16666145 Supplier Gilson™ Supplier No. 174732
  • ONLINE EXCLUSIVE PRICE
    1180.00 EUR
    992.92 EUR / Pack of 1
    Save 187.08 EUR
    15% Off
 Restricted Item
Estimated Shipment: 25-11-2026
These accessories, which include drop detector heads, dispense needles, collection plates, and a multiple column adapter, are for use with Gilson fraction collectors.
  • Purification Technique HPLC
  • Used With Fraction Collector
Specifications
Description HPLC Purification Technique
For Use With (Equipment) Fraction Collector