Learn More
Cole Parmer™ Multi-Roll Tape Dispenser
Product Code. 15273953 Shop All Cole Parmer Products
Change view
Click to view available options
Width (English):
9.5 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
15273953 9.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15273953 Supplier Cole Parmer™ Supplier No. 0653011
  • ONLINE EXCLUSIVE PRICE
    80.05 EUR
    69.06 EUR / Pack of 1
    Save 10.99 EUR
    13% Off
Estimated Shipment: 10-12-2026
Stores, carries, and cuts multiple tape rolls.
  • Holds single or multiple rolls of tape up to 9 1/2 inches wide.
  • Sturdy metal dispenser.
  • Accommodates a variety of different-sized tape.
  • Divided into four individual sections so that tape can be easily refilled within each section.
Specifications
Width (English) 9.5 in.
Material Metal
No. of Compartments 4
For Use With (Application) Tape dispenser
For Use With (Equipment) Tape rolls
Quantity 1 Ea.