Learn More
Miele Professional™ Mobile Injector Unit E 355
Product Code. 13112290 Shop All Miele Products
Change view
Click to view available options
For Use With (Equipment):
G 7804 and G 7883, Narrow-necked laboratory glassware/flasks/volumetrics
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13112290 G 7804 and G 7883, Narrow-necked laboratory glassware/flasks/volumetrics Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13112290 Supplier Miele Professional™ Supplier No. 3809560
  • ONLINE EXCLUSIVE PRICE
    2125.00 EUR
    1779.41 EUR / Pack of 1
    Save 345.59 EUR
    16% Off
Estimated Shipment: 11-01-2027
Mobile unit E 355 for the optimum loading of narrow necked glassware
  • equipped with 16 injector nozzles
  • Half can be equipped with other inserts
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-purpose Mobile Injector
For Use With (Equipment) G 7804 and G 7883, Narrow-necked laboratory glassware/flasks/volumetrics
Includes 16 Jets/clips