Learn More
Multi 7 ions, 10mg/L of each Component, for Ion Chromatography, H2O matrix, Fisher chemical™
BIOPHARMA
Product Code. 12932235 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12932235 100 mL 100mL
12942235 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12932235 Supplier Fisher Chemical Supplier No. A 101 100
  • ONLINE EXCLUSIVE PRICE
    174.00 EUR
    126.93 EUR / 100mL
    Save 47.07 EUR
    27% Off
 Restricted Item
Estimated Shipment: 05-01-2027
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Ions (7 components): F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
Specifications
Chemical Name or Material Multi 7 ions
Packaging Bottle
Analyte or Component Names F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
Physical Form Liquid
Concentration 10 mg/L of each Component
Type Multi-component Standard Solution
Element F-, Cl-, NO2-, Br-, NO23-, PO43-, SO42
Solvent or Matrix Water
Quantity 100 mL