Learn More
MTS PHOTO-SWITCHABLE Compound Hydrochloric Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | MTS Photo-Switchable Compound Hydrochloric Acid Salt |
| CAS | 2470235-57-3 |
| Molecular Formula | C31H39N5O5S2 . HCl |
| Quantity | 10 mg |
| Synonym | S-(3-((4-((4-(2-((4-Hydroxy-3,5-diisopropylbenzyl)amino)acetamido)phenyl)diazenyl)phenyl)amino)-3-oxopropyl) Methanesulfonothioate Hydrochloride Salt, MAP 20 Hydrochloride Salt |
| InChI Formula | InChI=1S/C31H39N5O5S2.ClH/c1-20(2)27-16-22(17-28(21(3)4)31(27)39)18-32-19-30(38)34-24-8-12-26(13-9-24)36-35-25-10-6-23(7-11-25)33-29(37)14-15-42-43(5,40)41;/h6-13,16-17,20-21,32,39H,14-15,18-19H2,1-5H3,(H,33,37)(H,34,38);1H |
| SMILES | O=C(NC1=CC=C(N=NC2=CC=C(NC(CNCC3=CC(C(C)C)=C(O)C(C(C)C)=C3)=O)C=C2)C=C1)CCSS(C)(=O)=O.Cl |
| IUPAC Name | S-(3-((4-((4-(2-((4-hydroxy-3,5-diisopropylbenzyl)amino)acetamido)phenyl)diazenyl)phenyl)amino)-3-oxopropyl) methanesulfonothioate hydrochloride |
| Molecular Weight (g/mol) | 662.26 |
| Formula Weight | 661.22 |