missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTPAP Polyclonal Antibody
GREENER_CHOICE

Product Code. 13455827
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13455827 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13455827 Supplier Invitrogen™ Supplier No. PA531312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116, A549, H1299, HCT116. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the DNA polymerase type-B-like family. This enzyme synthesizes the 3' poly(A) tail of mitochondrial transcripts and plays a role in replication-dependent histone mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTPAP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene MTPAP
Gene Accession No. Q9NVV4
Gene Alias FLJ10486; mitochondrial poly(A) polymerase; mtPAP; PAP; PAP-associated domain-containing protein 1; PAPD1; Poly(A) RNA polymerase, mitochondrial; polynucleotide adenylyltransferase; RP11305E6.3; RP11-305E6.3; SPAX4; terminal uridylyltransferase 1; TUTase 1
Gene Symbols MTPAP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human mtPAP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.