missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MTEP hydrochloride

Product Code. 18084392
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084392

Brand: Tocris Bioscience™ 2921/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mGlu5 antagonist

MTEP hydrochloride is a potent, selective and non-competitive mGlu5 antagonist (IC50 = 5 nM in Ca2+-flux assay; Ki= 16 nM). Displays anxiolytic activity in vivo and is orally active.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-((2-Methyl-1,3-thiazol-4-yl)ethynyl)pyridine hydrochloride
CAS 1186195-60-7
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100 mM in DMSO
Molecular Formula C11H8N2S.HCl
Formula Weight Observed MW: 236.72

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.