missing translation for 'onlineSavingsMsg'
Learn More

MTCP1NB Antibody [HRP], Novus Biologicals™

Product Code. 30757506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30757506 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30757506 Supplier Novus Biologicals Supplier No. NBP347010H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTCP1NB Polyclonal antibody specifically detects MTCP1NB in Human,Mouse samples. It is validated for ELISA,Serum
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCP1NB
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias C6.1B, mature T-cell proliferation 1, mature T-cell proliferation 1 neighbor, mature T-cell proliferation 1 neighbor protein, Mature T-cell proliferation-1 type A, MGC2069, MTCP1, MTCP-1 type A, p8, p8MTCP1, Protein p8 MTCP-1
Host Species Rabbit
Immunogen mature T-cell proliferation 1 neighbor. (Uniprot# P56277)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100272147
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.