missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/RPS6KA5 Antibody (PCRP-RPS6KA5-1A8), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | MSK1/RPS6KA5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PCRP-RPS6KA5-1A8 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human MSK1/RPS6KA5 protein (Uniprot: O75582) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?