missing translation for 'onlineSavingsMsg'
Learn More

MSC2530818, MedChemExpress

Product Code. 16464656
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464656 100 mg 100mg
16463458 10 mg 10mg
16439956 50 mg 50mg
16416187 5 mg 5mg
16412776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464656 Supplier Medchem Express Supplier No. HY101611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MSC2530818 is a potent, selective and orally available CDK8 inhibitor with an IC50 of 2.6 nM for CDK8.

MSC2530818 is a potent, selective and orally available CDK8 inhibitor with an IC50 of 2.6 nM for CDK8.

Specifications

Chemical Name or Material MSC2530818
CAS 1883423-59-3
Color White
Purity Grade Notes Research
Molecular Formula C18H17ClN4O
Quantity 100 mg
Solubility Information DMSO : 110 mg/mL (322.76 mM; Need ultrasonic)
SMILES O=C(N1[C@H](C2=CC=C(Cl)C=C2)CCC1)C3=CN=C(NN=C4C)C4=C3
Molecular Weight (g/mol) 340.81
Formula Weight 340.81
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.