missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A6A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | MS4A6A |
|---|---|
| Concentration | 0.45 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16839323
|
Proteintech
11930-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16829323
|
Proteintech
11930-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be involved in signal transduction as a component of a multimeric receptor complex.Specifications
| MS4A6A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H2W1 | |
| 64231 | |
| MS4A6A Fusion Protein Ag2464 | |
| Primary | |
| -20°C | |
| MS4A6A |
| 0.45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 4SPAN3, 4SPAN3.2, CD20 antigen like 3, CD20L3, CDA01, MS4A6, MS4A6A, MST090, MSTP090 | |
| MS4A6A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title