missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A6A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11930-1-AP-150UL
This item is not returnable.
View return policy
Description
May be involved in signal transduction as a component of a multimeric receptor complex.Specifications
| MS4A6A | |
| Polyclonal | |
| Unconjugated | |
| MS4A6A | |
| 4SPAN3, 4SPAN3.2, CD20 antigen like 3, CD20L3, CDA01, MS4A6, MS4A6A, MST090, MSTP090 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64231 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H2W1 | |
| MS4A6A | |
| MS4A6A Fusion Protein Ag2464 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction