missing translation for 'onlineSavingsMsg'
Learn More

MRPS17 Antibody [FITC], Novus Biologicals™

Product Code. 30749335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30749335 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30749335 Supplier Novus Biologicals Supplier No. NBP347025F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPS17 Polyclonal antibody specifically detects MRPS17 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS17
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias mitochondrial ribosomal protein S17, MRP-S1728S ribosomal protein S17, mitochondrial, RPMS17HSPC011, S17mt
Host Species Rabbit
Immunogen mitochondrial ribosomal protein S17. (Uniprot# Q9Y2R5)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51373
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.