missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL42 Monoclonal Antibody (3H6G11)
GREENER_CHOICE

Product Code. 15318070
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15318070 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15318070 Supplier Invitrogen™ Supplier No. MA517128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-17128 targets MRPL42 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17128 immunogen is purified recombinant fragment of human MRPL42 (amino acids: 142-203) expressed in E. Coli. MA5-17128 detects MRPL42 which has a predicted molecular weight of approximately 16.7kDa.

MRPL42 are mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a protein identified as belonging to both the 28S and the 39S subunits. Alternative splicing results in multiple transcript variants. Pseudogenes corresponding to this gene are found on chromosomes 4q, 6p, 6q, 7p, and 15q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL42
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3H6G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene MRPL42
Gene Accession No. Q9Y6G3
Gene Alias 2700009F22Rik; 28S ribosomal protein L42, mitochondrial; 28S ribosomal protein S32, mitochondrial; 2900055D03Rik; 39S ribosomal protein L31, mitochondrial; 39S ribosomal protein L42, mitochondrial; D10Ertd322e; HSPC204; L31mt; L42MT; mitochndrial ribosomal protein L42; Mitochondrial 28S ribosomal protein S32, mitochondrial; Mitochondrial large ribosomal subunit protein mL42; mitochondrial ribosomal protein L42; MRPL31; MRP-L31; MRPL42; MRP-L42; MRPS32; MRP-S32; PTD007; RPML31; S32mt
Gene Symbols MRPL42
Host Species Mouse
Immunogen Purified recombinant fragment of human MRPL42 (amino acids: 142-203) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.