Learn More
Tocris Bioscience™ MQAE
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
MQAE is a membrane-permeable intracellular chloride ion (Cl-) indicator. The fluorescence intensity of MQAE decreases proportionally as Cl- ions increase. Excitation/emission maxima (λ) are 355 nm and 460 nm respectively. Can be used in fluorescence microscopy and flow cytometry, suitable for use in vivo and in vitro.
Specifications
Specifications
| Chemical Name or Material | 1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide |
| CAS | 162558-52-3 |
| Quantity | 100 mg |
| Target | Ion Indicators |
| Molecular Formula | C14H16BrNO3 |
| Purity | 0.95 |