missing translation for 'onlineSavingsMsg'
Learn More

MPZL3 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30762378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762378 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762378 Supplier Novus Biologicals Supplier No. NBP347055CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MPZL3 Polyclonal antibody specifically detects MPZL3 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPZL3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias myelin protein zero-like 3, myelin protein zero-like protein 3
Host Species Rabbit
Immunogen myelin protein zero-like 3. (Uniprot# Q6UWV2)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196264
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.