Learn More
MP Biomedicals™ Yeast RPM Plasmid Isolation Kit
Product Code. 11363149 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Samples:
100 Preps
Unit Size:
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples Unit Size
11363149 100 Preps 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11363149 Supplier MP Biomedicals™ Supplier No. 112069400
  • ONLINE EXCLUSIVE PRICE
    695.00 EUR
    661.50 EUR / Pack of 100
    Save 33.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Yeast Rapid Pure Miniprep Plasmid Isolation Kit designed for yeast cells. Can isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).
  • The plasmid isolated from yeast is suitable for PCR and bacterial transformation.
  • The same kit can be used to isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).
  • Plasmid DNA mini-preps from yeast rescue positive prey plasmids after a yeast two-hybrid or a yeast one-hybrid screen.
Specifications
Sample Type Yeast Plasmid Prep
For Use With (Application) For PCR and bacterial transformation
Includes Yeast lysing matrix, Alkaline Lysis solution, neutralizing solution, spin Buffer, wash solution concentrate, spin filters, catch tubes, Pre-lysis Buffer
No. of Samples 100 Preps
Sample Volume 1.5 mL
Sample Source Yeast Culture
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.