Learn More
MP Biomedicals™ Wheat Germ, Powder
Product Code. 11495712 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2.26 kg
11.33 kg
45.35 kg
Unit Size:
100lb
25lb
5lb
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495712 11.33 kg 25lb
11405722 45.35 kg 100lb
11485712 2.26 kg 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11495712 Supplier MP Biomedicals™ Supplier No. 0290328825
  • ONLINE EXCLUSIVE PRICE
    140.00 EUR
    133.35 EUR / 25lb
    Save 6.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Food supplement
Description:
MP Wheat Germ is raw and has not been heat treated in any manner. Contains wheat germ, bran and middlings.
Physical Description:
Brown flakes/powder
Specifications
Product Type Animal Diet
Format Flakes
For Use With (Application) In molecular biology, wheat germ extract is used to carry out in-vitro translation experiments, as the plant embryo contains all the macromolecular components necessary for translating mRNA into amino Acids, but contains relatively low levels of its own mRNA
Quantity 11.33 kg
Storage Requirements 15°C to 30°C