Learn More
MP Biomedicals™ Trypsin, 2.5% (w/v) Solution
Product Code. 11484805 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484805 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11484805 Supplier MP Biomedicals™ Supplier No. 091689349
Porcine pancreas-derived enzyme commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues
  • The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.
  • One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell counting, viability and cryopreservation
pH Range 5 to 6
Content And Storage 0°C and -20°C
Form Solution
Quantity 100 mL
Product Type Trypsin