missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pokeweed Mitogen, Lyoph.

Product Code. 11455116 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1mg
5mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455116 1mg 1mg
11494176 5mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455116 Supplier MP Biomedicals™ Supplier No. 08780332
  • ONLINE EXCLUSIVE PRICE
    167.00 EUR
    158.55 EUR / 1mg
    Save 8.45 EUR
    5% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phytolacca Americana

Specifications

For Use With (Application) Immunoassay
Name Pokeweed Mitogen
pH Range 7.2 to 7.4
Quantity 1mg
Storage Requirements 2° to 8°C
Recombinant Recombinant
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.