missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.
Specifications
Specifications
| For Use With (Application) | Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies |
| pH Range | 1 to 3.5 |
| Sample Type | Porcine Stomach Mucosa |
| Content And Storage | 0°C |
| Form | Lyophilized |
| Quantity | 5 g |
| Product Type | Pepsin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?