missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pancreatin 8X, USP Grade

Product Code. 11416381 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11416381

Brand: MP Biomedicals™ 0219397680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From porcine pancreas

Composed of numerous enzymes including amylase, trypsin, lipase, ribonuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in five minutes in water at 40°C, and not less than than 25 times its weight of casein into proteoses. It hydrolyzes fats to glycerol and fatty acid, protein to proteoses and derived compounds, and starch into dextrins and sugar. Lipase Activity: ≥16U/mg. Amylase Activity: ≥200U/mg. Protease Activity: ≥200U/mg.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 8X
Color Cream
For Use With (Application) Promote esophageal tissue loosening in patients with esophageal tumors
Content And Storage 0°C
Source Porcine pancreas
Quantity 100 g
Product Type Pancreatin
Form Powder
compliance-icons compliance-icons
Product Identifier
  • Pancreatin 8×, USP
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Pancreatin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.