Learn More
MP Biomedicals™ Pancreatin 8X, USP Grade
Shop All MP Biomedicals ProductsDescription
Composed of numerous enzymes including amylase, trypsin, lipase, ribonuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in five minutes in water at 40°C, and not less than than 25 times its weight of casein into proteoses. It hydrolyzes fats to glycerol and fatty acid, protein to proteoses and derived compounds, and starch into dextrins and sugar. Lipase Activity: ≥16U/mg. Amylase Activity: ≥200U/mg. Protease Activity: ≥200U/mg.
Specifications
Specifications
| Concentration | 8X |
| Color | Cream |
| For Use With (Application) | Promote esophageal tissue loosening in patients with esophageal tumors |
| Content And Storage | 0°C |
| Source | Porcine pancreas |
| Quantity | 25 g |
| Product Type | Pancreatin |
| Form | Powder |
Safety and Handling
- Pancreatin, USP
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : pancreatin
Your input is important to us. Please complete this form to provide feedback related to the content on this product.