missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Menhaden Oil

Product Code. 11452831 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 gal.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452831 1 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452831 Supplier MP Biomedicals™ Supplier No. 0296012001
  • ONLINE EXCLUSIVE PRICE
    147.00 EUR
    139.65 EUR / Pack of 1
    Save 7.35 EUR
    5% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Menhaden Oil, refined

Menhaden oil is use as a control or to check sample for fatty acid methyl ester (FAME) analyses. It is rich in omega-3 fatty acids. When used to partially supplement a high-fat diet in diabetic rats, Menhaden oil improved end points for peripheral neuropathy by improving motor and sensory nerve conduction velocity. Decreased accumulation of fatty acids in rat livers was also demonstrated.

Specifications

Product Type Animal Diet
Format Lyophilized
For Use With (Application) Menhaden Oil is use as a control or check sample for Fatty Acid Methyl Ester (FAME) analyses
Quantity 1 gal.
Storage Requirements -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.