missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysozyme from Chicken Egg White, 20,000U/mg
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
Description
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS Number: 9001-63-2
- EC Number: 232-620-4
- MDL Number: MFCD00131557
- Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
- Specific Activity: ≤20,000U/mg solid
- Extinction Coefficient (E1%): 26.4 (280 nm)
- Isoelectric point (pI): pH 11.0
- pH: 3.6
Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.
Specifications
Specifications
| Concentration | 3X |
| Color | White |
| pH | 11 |
| For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
| Content And Storage | 0°C |
| Source | Chicken egg white |
| Quantity | 1 g |
| Product Type | Lysozyme |
| Molecular Weight (g/mol) | 14388 |
| Inhibitors | Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
| Show More |
Safety and Handling
Product Identifier
- Lysozyme
- Warning
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : lysozyme
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction