Learn More
MP Biomedicals™ Lotus tetragonolobus Agglutinin
Product Code. 11438701 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438701 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11438701 Supplier MP Biomedicals™ Supplier No. 0215330305
  • ONLINE EXCLUSIVE PRICE
    106.00 EUR
    98.75 EUR / 5mg
    Save 7.25 EUR
    6% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Exhibits a specificity for α-L-fucose.
Specifications
Quantity 5 mg
Content And Storage -20°C
Form Powder
Product Type Lotus tetragonolobus
Specificity L-fucose