missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ β-Glucosidase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 kU
25 kU
5 kU
Unit Size:
100ku
25ku
5ku
Description
Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.
Specifications
Specifications
| Concentration | ≥1000 U/mg |
| Color | White |
| For Use With (Application) | Synthesis of glucosides and fucosides |
| Content And Storage | -20°C |
| Source | Sweet almonds |
| Quantity | 100 kU |
| Product Type | beta-Glucosidase |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction