missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | >16000 U/g |
| Color | Tan |
| pH | 4.2 to 5.2 |
| For Use With (Application) | Molecular biology, cell biology |
| Content And Storage | 0°C |
| Source | Trichoderma viride |
| Quantity | 5 g |
| Product Type | Cellulase |
| Molecular Weight (g/mol) | C1 57000, 42000, Endoglucanase 52000, 23500 to 58000, Exoglucanase 60500 to 62000, b-Glucosidase 76000 |
| Form | Powder |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?