missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Activity 2000units/mg
Specifications
Specifications
| Concentration | >2000 U/mg |
| Color | Tan |
| For Use With (Application) | Molecular biology, cell biology |
| Content And Storage | 2°C to 8°C |
| Source | Bovine erythrocytes |
| Quantity | 100 mg |
| Product Type | Carbonic Anhydrase |
| Molecular Weight (g/mol) | 30000 |
| Form | Lyophilized Powder |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?