missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ beta-Xylosidase from Trichoderma viride

Product Code. 11426901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 U
Unit Size:
5 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426901 5 U 5 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11426901 Supplier MP Biomedicals™ Supplier No. 0219893201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For glucose or glycogen metabolism

  • Synonyms: 1,4-β-Xylan xylohydrolase
  • CAS Number: 9025-53-0
  • EC Number: 232-797-8
  • Available as lyophilized
  • Activity: 5U/vial

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/Vial
pH 4
For Use With (Application) Cell biology and analysis, Glucose/Glgycogen metabolism
Content And Storage 2°C to 8°C
Source Trichoderma viride
Quantity 5 U
Product Type beta-Xylosidase
Form Lyophilized Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.