missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Avidin-HRP

Product Code. 11424255 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424255 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424255 Supplier MP Biomedicals™ Supplier No. 0855898

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for use as a secondary reagent with biotin-labeled antibodies, antigens or probes in assays or staining of blots, cell suspensions or tissue sections

TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
pH 7.3
Quantity 5 mg
Content And Storage 2°C to 8°C
Form Liquid
For Use With (Application) Immunoassay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.