Learn More
MP Biomedicals™ Ascorbate Oxidase
From Cucumis species
Brand: MP Biomedicals™ 0219007582
Additional Details : CAS Number : 9029-44-1 Weight : 0.01000kg
Description
Activity: 200U/mg solid. Unit Definition: One unit will oxidize 1.0μmole of L-ascorbate to dehydroascorbate per minute at pH 5.6 and20 °C. Lyophilized powder.
Specifications
≥200 U/mg | |
7 to 10 | |
0°C | |
200 U | |
1320001, 140000 | |
Lyophilized Powder |
Light Blue | |
Enzymatic determination of Ascorbic Acid | |
Cucumis species | |
Ascorbate Oxidase | |
Azide, Cyanide, Na2S, Diethyldithiocarbamate (Na) |