Learn More
MP Biomedicals™ Antibiotics: Tetracycline hydrochloride
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Brand: MP Biomedicals™ 0219454201_CLEAN
Description
- Format: Crystalline Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.