missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ alpha-Chymotrypsin from Bovine Pancreas

Product Code. 11438521 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11438521

Brand: MP Biomedicals™ 02152272.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on carboxyl end of the bond

α-Chymotrypsin preferentially catalyzes the hydrolysis of peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan.

  • CAS Number: 9004-07-3
  • EC Number: 232-671-2
  • MDL Number: MFCD00130481
  • Extinction Coefficient: 20.4
  • Format: white powder
  • From Bovine Pancreas
  • Crystallized, dialyzed, salt-free lyophilized powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Color White
For Use With (Application) Catalyzes the hydrolysis of Peptide bonds involving L-isomers of Tyrosine, Phenylalanine, Tryptophan
Content And Storage 0°C
Source Bovine pancreas
Quantity 250 mg
Product Type alpha-Chymotrypsin
Molecular Weight (g/mol) 25
Inhibitors Heavy Metals, natural Trypsin inhibitors to various degrees, Organophosphorus compounds, AEBSF, alpha-1-antitrypsin, Aprotinin, DFP, PMSF, TPCK and a-2-Macroglobulin
Form Crystallized, Lyophilized
compliance-icons compliance-icons
Product Identifier
  • Alpha-Chymotrypsin, 1× Crystallized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Chymotrypsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.