missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Alkaline Phosphatase, Powder

Product Code. 11476091 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
10 kU
100 kU
Unit Size:
100ku
10ku
1ku
This item is not returnable. View return policy

Product Code. 11476091

Brand: MP Biomedicals™ 0219068010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From calf intestine

Activity: ∽1.0U/mg. Unit Definition: One unit causes hydrolysis of one micromole of p-nitrophenyl phosphate per minute at pH 9.6 and 25°C. Freeze-dried powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1 U/mg
Color Off-White
pH 9.8
For Use With (Application) Used to remove 5'-phosphate groups from Nucleic Acid molecules to prevent self-ligation of DNA fragments and vectors during recombinant DNA manipulation
Content And Storage -20°C
Source Calf intestine
Quantity 10 kU
Product Type Alkaline Phosphatase
Molecular Weight (g/mol) 140000
Inhibitors Acidification to 4.5 pH reversibly inactives the Enzyme, Inorganic Phosphate, chelating agents, EDTA
Form Lyophilized Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.