Learn More
MP Biomedicals™ AIN-93-Vx Vitamin Mix
Product Code. 11431452 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431452 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11431452 Supplier MP Biomedicals™ Supplier No. 0296040201
  • ONLINE EXCLUSIVE PRICE
    264.00 EUR
    250.95 EUR / 1kg
    Save 13.05 EUR
    4% Off
 Restricted Item
In Stock
Component of AIN-93M & AIN-93G diet
AIN-93-VX Vitamin mix is a component of AIN-93M & AIN-93 g diet.
Specifications
Product Type Animal Diet
Format Powder
Quantity 1 kg
Species Rodent
Storage Requirements 15°C to 30°C