missing translation for 'onlineSavingsMsg'
Learn More

Mozavaptan, TRC

Product Code. 30135299
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135299 5 mg 5mg
30008533 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135299 Supplier Toronto Research Chemicals Supplier No. TRCM7327205MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137975-06-5
Molecular Formula C27H29N3O2
Molecular Weight (g/mol) 427.54
Synonym N-[4-[[5-(Dimethylamino)-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]carbonyl]phenyl]-2-methylbenzamide, OPC 31260;
IUPAC Name N-[4-[5-(dimethylamino)-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl]phenyl]-2-methyl-benzamide
SMILES CN(C)C1CCCN(C(=O)c2ccc(NC(=O)c3ccccc3C)cc2)c4ccccc14

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H29N3O2/c1-19-9-4-5-10-22(19)26(31)28-21-16-14-20(15-17-21)27(32)30-18-8-13-24(29(2)3)23-11-6-7-12-25(23)30/h4-7,9-12,14-17,24H,8,13,18H2,1-3H3,(H,28,31)
Formula Weight 427.226
Chemical Name or Material Mozavaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.