Learn More
Mozavaptan, MedChemExpress
Product Code. 16473527
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473527 50 mg 50mg
16447508 10 mg 10mg
16427466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16473527 Supplier Medchem Express Supplier No. HY18346
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Mozavaptan (OPC-31260) is a benzazepine derivative and a potent, selective, competitive and orally active vasopressin V2 receptor antagonist with an IC50 of 14 nM.
Mozavaptan shows ∼85-fold selectivity for V2 receptor over V1 receptor (IC50 of 1.2 μM), and can antagonize the antidiuretic action of arginine vasopressin (AVP) in vivo. Mozavaptan has the potential for hyponatremia, syndrome of inappropriate antidiuretic hormone (SIADH), and congestive heart failure treatment.
Specifications
Chemical Name or Material Mozavaptan
Physical Form Solid
CAS 137975-06-5
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H29N3O2
For Use With (Application) Cancer-programmed cell death
Synonym OPC-31260
Solubility Information DMSO : 6.2 mg/mL (14.50 mM; Need warming)
SMILES CC1=CC=CC=C1C(NC2=CC=C(C(N3C(C=CC=C4)=C4C(N(C)C)CCC3)=O)C=C2)=O
Molecular Weight (g/mol) 427.54
Formula Weight 427.54
Percent Purity 98.12%
Grade Research