missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MGL1/2 (CD301a/b) Antibody, R&D Systems™
Description
MGL1/2 (CD301a/b) Polyclonal specifically detects MGL1/2 (CD301a/b) in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
Specifications
| Antigen | MGL1/2 (CD301a/b) |
| Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Gene Accession No. | AAH14811 |
| Gene Alias | ASGP-BP, CD301a, CD301b, CD8, Leu2, p32 |
| Host Species | Goat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse MGL1 Gln57-Ser304 Accession # AAH14811 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?